Learn More
Trajan Scientific Liner Inlet Liner Straight PerkinElmer 5/PK
SDP
Catalog No. 5000494909
Encompass_Preferred
SGE PerkinElmer straight inlet liners 2 mm ID length 92 mm PK5
Catalog No. 50-004-94909 Supplier Trajan Scientific Supplier No. 092103
SGE PerkinElmer straight inlet liners 2 mm ID length 92 mm PK5