Learn More
Trajan Scientific Liner Inlet Liner Taper f.e.l.t. 1/PK
SDP
Catalog No. 5000494583
Encompass_Preferred
SGE taper inlet liner with f e l t with pre-fitted CRS ONE o-ring 4 mm ID length 78 5 mm PK1
Catalog No. 50-004-94583 Supplier Trajan Scientific Supplier No. 09260001
SGE taper inlet liner with f e l t with pre-fitted CRS ONE o-ring 4 mm ID length 78 5 mm PK1