Learn More
Trajan Scientific Liner Inlet Liner Taper f.e.l.t. 5/PK
SDP
Catalog No. 5000494289
Encompass_Preferred
SGE taper inlet liners with f e l t with pre-fitted CRS ONE o-ring 4 mm ID length 78 5 mm PK5
Catalog No. 50-004-94289 Supplier Trajan Scientific Supplier No. 092600
SGE taper inlet liners with f e l t with pre-fitted CRS ONE o-ring 4 mm ID length 78 5 mm PK5