Learn More
Trajan Scientific Liner Inlet Liner Taper Fison 5/PK
SDP
Catalog No. 5000493662
Encompass_Preferred
SGE Thermo Scientific taper inlet liners 3 mm ID length 105 mm PK5
Catalog No. 50-004-93662 Supplier Trajan Scientific Supplier No. 092141
SGE Thermo Scientific taper inlet liners 3 mm ID length 105 mm PK5