Learn More
Trajan Scientific Liner Inlet Liner Taper Shimadzu 5/PK
SDP
Catalog No. 5000494605
Encompass_Preferred
SGE Shimadzu taper inlet liners 3 4 mm ID length 95 mm PK5
Catalog No. 50-004-94605 Supplier Trajan Scientific Supplier No. 092071
SGE Shimadzu taper inlet liners 3 4 mm ID length 95 mm PK5