Learn More
Trajan Scientific Liner Inlet Liner Varian Focus 5/PK
SDP
Catalog No. 5000494932
Encompass_Preferred
SGE Bruker/Varian FocusLiners 4 mm ID (top-end rest) length 72 mm PK5
Catalog No. 50-004-94932 Supplier Trajan Scientific Supplier No. 092028
SGE Bruker/Varian FocusLiners 4 mm ID (top-end rest) length 72 mm PK5