Learn More
Trajan Scientific Liner Inlet Liner Varian Medical 5/PK
SDP
Catalog No. 5000493773
Encompass_Preferred
SGE Bruker/Varian straight inlet liners 0 5 mm ID length 54 mm PK5
Catalog No. 50-004-93773 Supplier Trajan Scientific Supplier No. 092027
SGE Bruker/Varian straight inlet liners 0 5 mm ID length 54 mm PK5