Learn More
Trajan Scientific Liner Inlet Liner Varian Restriction 5/PK
SDP
Catalog No. 5000494887
Encompass_Preferred
SGE Bruker/Varian inlet liners restricted 2 mm ID length 74 mm PK5
Catalog No. 50-004-94887 Supplier Trajan Scientific Supplier No. 092024
SGE Bruker/Varian inlet liners restricted 2 mm ID length 74 mm PK5