Learn More
Trajan Scientific Liner Inlet Liner Varian Restriction 5/PK
SDP
Supplier:  Trajan Scientific 092030
Encompass_Preferred
SGE Bruker/Varian straight inlet liners 0 8 mm ID (rest 0 25 mm) length 54 mm PK5
Catalog No. 50-004-94451