Learn More
Trajan Scientific Liner Inlet Liner Varian Straight 5/PK
SDP
Catalog No. 5000493586
Encompass_Preferred
SGE Bruker/Varian straight inlet liners 0 5 mm ID length 54 mm PK5
Catalog No. 50-004-93586 Supplier Trajan Scientific Supplier No. 092031
SGE Bruker/Varian straight inlet liners 0 5 mm ID length 54 mm PK5