Learn More
Trajan Scientific Liner Inlet Liner Varian Taper 5/PK
SDP
Catalog No. 5000493950
Encompass_Preferred
SGE Bruker/Varian taper inlet liners 3 4 mm ID length 54 mm PK5
Catalog No. 50-004-93950 Supplier Trajan Scientific Supplier No. 092038
SGE Bruker/Varian taper inlet liners 3 4 mm ID length 54 mm PK5