Learn More
Trajan Scientific Liner Inlet Liner Varian Taper 5/PK
SDP
Catalog No. 5000494346
Encompass_Preferred
SGE Bruker/Varian taper inlet liners 2 mm ID length 54 mm PK5
Catalog No. 50-004-94346 Supplier Trajan Scientific Supplier No. 092039
SGE Bruker/Varian taper inlet liners 2 mm ID length 54 mm PK5