Learn More
Chemglass Life Sciences Liner, PTFE, 5mm, NMR, Tube
SDP
Catalog No. CTL57
Encompass_Preferred
Liner, PTFE, 5mm, NMR, Tube. NORELL™ PTFE Liners have a thin-wall construction that minimizes filling-factor losses. They are supplied with a PTFE plug closure. These liners should be used for NMR studies where chemical compounds such as hydrofluoric acid, ammonium bifluroride and concentrated hydroxide solutions are present.C-TL-5-7: Volume at 50mm = ~ 0.35mL for the 5mm NMR Tube
Catalog No. CTL57 Supplier Chemglass Life Sciences Supplier No. CTL57
Liner, PTFE, 5mm, NMR, Tube. NORELL™ PTFE Liners have a thin-wall construction that minimizes filling-factor losses. They are supplied with a PTFE plug closure. These liners should be used for NMR studies where chemical compounds such as hydrofluoric acid, ammonium bifluroride and concentrated hydroxide solutions are present.C-TL-5-7: Volume at 50mm = ~ 0.35mL for the 5mm NMR Tube