Learn More
Trajan Scientific LINER SPLITLS SHIMAD 17A DEACT
SDP
Catalog No. 502530179
Encompass_Preferred
Trajan Shimadzu Liner, Quantity: 5/Pk., Diameter Inner: 3.4 mm, Diameter Outer: 5 mm, Length: 95 mm, Split, Straight-Through
Catalog No. 50-253-0179 Supplier Trajan Scientific Supplier No. 092064
Trajan Shimadzu Liner, Quantity: 5/Pk., Diameter Inner: 3.4 mm, Diameter Outer: 5 mm, Length: 95 mm, Split, Straight-Through