Learn More
Trajan Scientific LINER SPLT F/HP5890 4MM DEACT
SDP
Catalog No. 502530187
Encompass_Preferred
SGE Split Liner With Quartz Wool, Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Packing Material: Quartz Wool, GC Split Liner, Quantity: 5/Pk.
Catalog No. 50-253-0187 Supplier Trajan Scientific Supplier No. 092001
SGE Split Liner With Quartz Wool, Diameter Inner: 0.4 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Packing Material: Quartz Wool, GC Split Liner, Quantity: 5/Pk.