Learn More
Med Vet International Dynarex Disposable Suction Liner, 1500 cc, 50/Case
Catalog No. 503042893
Encompass_Preferred
Dynarex Resp-02™ Disposable Suction Liners are designed to effectively and safely collect and contain aspirated fluids and materials. Each high-quality, durable liner features a patient-port elbow adaptor that allows quick and easy hookup and helps prevent tubing from kinking. The liner is ultrasonically sealed to the lid—so no assembly is needed. Suction liners require the use of a reusable outer canister. Ultrasonically welded shut-off valve helps prevent overflow and contamination Lid comes sealed and pre-assembled with pour spout, vacuum, and patient ports All port caps are engineered to create an air-tight snuggle seal Not made with natural rubber latex or DEHP
Catalog No. 50-304-2893 Supplier Med Vet International Supplier No. 32261
Dynarex Resp-02™ Disposable Suction Liners are designed to effectively and safely collect and contain aspirated fluids and materials. Each high-quality, durable liner features a patient-port elbow adaptor that allows quick and easy hookup and helps prevent tubing from kinking. The liner is ultrasonically sealed to the lid—so no assembly is needed. Suction liners require the use of a reusable outer canister. Ultrasonically welded shut-off valve helps prevent overflow and contamination Lid comes sealed and pre-assembled with pour spout, vacuum, and patient ports All port caps are engineered to create an air-tight snuggle seal Not made with natural rubber latex or DEHP