Learn More
Elkay Plastics Company Liner Linear Low Density Red Infectious Waste Print 2 Gallon Capacity 1.5 mil 11 X 14 500 per Roll
Catalog No. 501049775
Encompass_Preferred
These liners are stronger than their low density polyethylene counterparts. Linear low density liners come with star-sealed bottoms and protect the handler from its contents. All bags display the biohazard symbol and warnings in English and Spanish. Meets federal and state regulations for tear resistance of 480 grams (ASTM D1922) and impact resistance of 165 grams (ASTM D1709).
Catalog No. 50-104-9775 Supplier Elkay Plastics Company Supplier No. LBOR1114RE
These liners are stronger than their low density polyethylene counterparts. Linear low density liners come with star-sealed bottoms and protect the handler from its contents. All bags display the biohazard symbol and warnings in English and Spanish. Meets federal and state regulations for tear resistance of 480 grams (ASTM D1922) and impact resistance of 165 grams (ASTM D1709).