Learn More
Essendant Grease-Proof Quilon Pan Liners, 16.38 x 24.38, White, 1,000 Sheets/Carton
Catalog No. 2793709
Encompass_Preferred
Chromium-free, quilon surface treatment ensures easy release, without the need for grease and oils. Multi-use design helps minimize waste, helping you lower your operating costs. Flat-pack storage box.
Catalog No. 27-937-09 Supplier Essendant Supplier No. BGC030001
Chromium-free, quilon surface treatment ensures easy release, without the need for grease and oils. Multi-use design helps minimize waste, helping you lower your operating costs. Flat-pack storage box.