Learn More
CEM Corporation LINERS 35ML 10/PK
SDP
Catalog No. NC0759154
Encompass
35mL Teflon PFA liner for Quartz Vials, set of 10. Reusable liners for use with Quartz vials. Provides a Teflon PFA wetted surface. MUST BE USED WITH A QUARTZ VIAL. WILL NOT WORK WITH PYREX
Catalog No. NC0759154 Supplier CEM Corporation Supplier No. 171978
35mL Teflon PFA liner for Quartz Vials, set of 10. Reusable liners for use with Quartz vials. Provides a Teflon PFA wetted surface. MUST BE USED WITH A QUARTZ VIAL. WILL NOT WORK WITH PYREX