Learn More
Beckman Coulter Beckman Coulter LINERS STRLE, 1L, QTY 6
Catalog No. 501632000
Encompass_Preferred
Beckman Coulter HarvestLine system disposable sterile liners, 1L, used with the 1L bottles for the JLA-9.1000 and JLA-8.1000 fixed angle rotors. (Qty 6)
Catalog No. 50-163-2000 Supplier Beckman Coulter Supplier No. C15217
Beckman Coulter HarvestLine system disposable sterile liners, 1L, used with the 1L bottles for the JLA-9.1000 and JLA-8.1000 fixed angle rotors. (Qty 6)