Learn More
TARGETMOL CHEMICALS INC LINETASTINE 5MG
SDP
Catalog No. 502580740
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Linetastine (TMK-688) is a 5-lipoxygenase inhibitor (5-LOX) that inhibits leukotriene production and antagonizes the effects of histamine.Linetastine is used in studies of asthma atherosclerosis and peptic ulcers. purity: 99%
Catalog No. 50-258-0740 Supplier TARGETMOL CHEMICALS INC Supplier No. T257375MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Linetastine (TMK-688) is a 5-lipoxygenase inhibitor (5-LOX) that inhibits leukotriene production and antagonizes the effects of histamine.Linetastine is used in studies of asthma atherosclerosis and peptic ulcers. purity: 99%