Learn More
Medchemexpress LLC HY-10394 10mg Medchemexpress, Linezolid CAS:165800-03-3 Purity:>98%
SDP
Catalog No. 501932250
Encompass_Preferred
Medchemexpress, HY-10394 10mg Linezolid CAS:165800-03-3 Linezolid (PNU-100766) is a synthetic antibiotic used for the treatment of serious infections caused by Gram-positive bacteria that are resistant to several other antibiotics. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2250 Supplier Medchemexpress LLC Supplier No. HY1039410MG
Medchemexpress, HY-10394 10mg Linezolid CAS:165800-03-3 Linezolid (PNU-100766) is a synthetic antibiotic used for the treatment of serious infections caused by Gram-positive bacteria that are resistant to several other antibiotics. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.