Learn More
Apexbio Technology LLC Linezolid(Synonyms: Zyvox, Zyvoxid, U-100766, PNU-100766, U-100766E, PNU100766, Linezolide), 10mM (in 1mL DMSO), CAS: 165800-03-3.
SDP
Catalog No. 5000060189
Encompass_Preferred
Linezolid (CAS 165800-03-3) is a synthetic oxazolidinone antimicrobial compound with broad-spectrum activity primarily against Gram-positive bacteria including multi-drug resistant pathogens such as methicillin-resistant Staphylococcus aureus (MRSA) vancomycin-resistant Enterococcus (VRE) and penicillin-resistant Streptococcus pneumoniae Linezolid inhibits bacterial protein synthesis by binding to the 23S ribosomal RNA of the 50S subunit thereby preventing formation of the functional 70S initiation complex In cell-free transcription-translation assays using E coli linezolid demonstrates an IC50 of approximately 1 8 mM Due to its pharmacokinetics and high oral bioavailability linezolid is widely utilized in clinical research related to complicated skin infections and bacterial pneumonia
Catalog No. 50-000-60189 Supplier Apexbio Technology LLC Supplier No. A51815.1
Linezolid (CAS 165800-03-3) is a synthetic oxazolidinone antimicrobial compound with broad-spectrum activity primarily against Gram-positive bacteria including multi-drug resistant pathogens such as methicillin-resistant Staphylococcus aureus (MRSA) vancomycin-resistant Enterococcus (VRE) and penicillin-resistant Streptococcus pneumoniae Linezolid inhibits bacterial protein synthesis by binding to the 23S ribosomal RNA of the 50S subunit thereby preventing formation of the functional 70S initiation complex In cell-free transcription-translation assays using E coli linezolid demonstrates an IC50 of approximately 1 8 mM Due to its pharmacokinetics and high oral bioavailability linezolid is widely utilized in clinical research related to complicated skin infections and bacterial pneumonia