Learn More
Medchemexpress LLC Linezolid | 165800-03-3 | MFCD00937825 | 100.0% | 337.35 g/mol | C16H20FN3O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY10394R5MG
Encompass_Preferred
Linezolid standard is an analytical reference material of the oxazolidinone antibiotic provided as a high-purity solid for use in method development, calibration, and quantitative analysis in research laboratories.
  • High purity of 99.99% for accurate calibration and quantitation
  • Supplied as a 5 mg pack suitable for small-scale analytical use
  • Compatible with HPLC and LC-MS workflows for method development
  • Chemical formula C16H20FN3O4 and molecular weight 337.35 g/mol
  • Accompanied by SDS and product documentation for handling and traceability
Catalog No. 50-002-43564