Learn More
AdipoGen Linezolid
SDP
Catalog No. 502560605
Encompass_Preferred
Chemical. CAS 165800-03-3. Formula C16H20FN3O4. MW 337.35. Synthetic. Linezolid is a synthetic oxazolidinone antibiotic with activity against a wide range of Gram-positive bacteria. Displays potent antibacterial activity against a variety of multidrug-resistant gram-positive microbes in vitro and in vivo, including resistant strains of several species, such as methicillin-resistant S. aureus MRSA, penicillin-resistant pneumococci, vancomycin-resistant enterococci, various anaerobic bacteria, and several mycobacteria and streptococci MICs = 2-4mg/ml. Linezolid inhibits protein synthesis by binding to domain V of the 23S ribosomal RNA of the 50S subunit of bacterial ribosomes and preventing the formation of a functional 70S initiation complex, thus inhibiting bacterial mRNA translation. Linezolid is also a weak, reversible, nonselective inhibitor of monoamine oxidase.
Catalog No. 50-256-0605 Supplier AdipoGen Supplier No. CDXL0031M025
Chemical. CAS 165800-03-3. Formula C16H20FN3O4. MW 337.35. Synthetic. Linezolid is a synthetic oxazolidinone antibiotic with activity against a wide range of Gram-positive bacteria. Displays potent antibacterial activity against a variety of multidrug-resistant gram-positive microbes in vitro and in vivo, including resistant strains of several species, such as methicillin-resistant S. aureus MRSA, penicillin-resistant pneumococci, vancomycin-resistant enterococci, various anaerobic bacteria, and several mycobacteria and streptococci MICs = 2-4mg/ml. Linezolid inhibits protein synthesis by binding to domain V of the 23S ribosomal RNA of the 50S subunit of bacterial ribosomes and preventing the formation of a functional 70S initiation complex, thus inhibiting bacterial mRNA translation. Linezolid is also a weak, reversible, nonselective inhibitor of monoamine oxidase.