Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linezolid-d3 (PNU-100766-d3) | 1127120-38-0 | MFCD08063578 | >99.9% | 340.36 g/mol | C16H17D3FN3O4 | 1 MG
SDP

Catalog No. 5000241869
Encompass_Preferred

Linezolid-d3 (PNU-100766-d3) is a deuterium-labeled analogue of the antibiotic linezolid designed for use as an isotopic internal standard and analytical reference in research and bioanalysis. It provides a defined mass shift for accurate quantitation by LC-MS/MS and other mass spectrometry methods.

  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High chemical and isotopic purity (>99.9% reported).
  • Molecular formula C16H17D3FN3O4 and molecular weight 340.36 g/mol.
  • Available in milligram quantities suitable for method development and validation.
  • Supplied as a neat solid for solution preparation and spiking workflows.

Catalog No. 50-002-41869 Supplier Medchemexpress LLC Supplier No. HY10394S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linezolid-d3 (PNU-100766-d3) is a deuterium-labeled analogue of the antibiotic linezolid designed for use as an isotopic internal standard and analytical reference in research and bioanalysis. It provides a defined mass shift for accurate quantitation by LC-MS/MS and other mass spectrometry methods.

  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High chemical and isotopic purity (>99.9% reported).
  • Molecular formula C16H17D3FN3O4 and molecular weight 340.36 g/mol.
  • Available in milligram quantities suitable for method development and validation.
  • Supplied as a neat solid for solution preparation and spiking workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.