Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linezolid (standard) | 165800-03-3 | MFCD00937825 | 100.0% | 337.35 g/mol | C16H20FN3O4 | 25 MG
SDP

Catalog No. 5000243312
Encompass_Preferred

Linezolid (Standard) is an analytical reference standard of the antibiotic linezolid intended for research and analytical use. It is provided as a 25 mg solid for qualitative and quantitative assays, method development, and verification in HPLC, GC, and MS.

  • Analytical reference standard for method development and validation.
  • High purity (99.99% by HPLC) for accurate assay calibration.
  • Suitable for HPLC, GC, and MS analyses.
  • Molecular formula C16H20FN3O4; molecular weight 337.35 g/mol.
  • Provided as a small solid quantity for precise weighing and storage.

Catalog No. 50-002-43312 Supplier Medchemexpress LLC Supplier No. HY10394R25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linezolid (Standard) is an analytical reference standard of the antibiotic linezolid intended for research and analytical use. It is provided as a 25 mg solid for qualitative and quantitative assays, method development, and verification in HPLC, GC, and MS.

  • Analytical reference standard for method development and validation.
  • High purity (99.99% by HPLC) for accurate assay calibration.
  • Suitable for HPLC, GC, and MS analyses.
  • Molecular formula C16H20FN3O4; molecular weight 337.35 g/mol.
  • Provided as a small solid quantity for precise weighing and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.