Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linezolid (standard) | 165800-03-3 | MFCD00937825 | 100.0% | 337.35 g/mol | C16H20FN3O4 | 50 MG
SDP

Catalog No. 5000244142
Encompass_Preferred

Linezolid (Standard) is an analytical reference standard of the antibiotic linezolid intended for research and analytical applications. It is provided as a high-purity material for use in method development, calibration, and quality control.

  • Purity 99.99%.
  • CAS number 165800-03-3.
  • Supplied in multiple package sizes, including 50 mg.
  • Intended for analytical method development, calibration, and quality control.
  • Certificate of analysis and safety data sheet available.
  • Molecular weight 337.35 g/mol; formula C16H20FN3O4.

Catalog No. 50-002-44142 Supplier Medchemexpress LLC Supplier No. HY10394R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linezolid (Standard) is an analytical reference standard of the antibiotic linezolid intended for research and analytical applications. It is provided as a high-purity material for use in method development, calibration, and quality control.

  • Purity 99.99%.
  • CAS number 165800-03-3.
  • Supplied in multiple package sizes, including 50 mg.
  • Intended for analytical method development, calibration, and quality control.
  • Certificate of analysis and safety data sheet available.
  • Molecular weight 337.35 g/mol; formula C16H20FN3O4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.