Learn More
Aviva Systems Biology Corp LINGO4 Antibody - middle region (ARP44480_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44480_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LINGO4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LINGO4. Alternate names: DAAT9248, LRRN6D, PRO34002ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58823