Learn More
Medchemexpress LLC Linifanib | 796967-16-3 | 99.4% | C21H18FN5O | 200 MG
SDP
Catalog No. 5000358080
Encompass_Preferred
Linifanib (ABT-869) is a potent, orally active, multi-target inhibitor of the VEGFR and PDGFR family. It demonstrates prominent antitumor activity and has significantly less activity against unrelated RTKs, soluble tyrosine kinases, or serine/threonine kinases. Linifanib also acts as a specific miR-10b inhibitor, blocking miR-10b biogenesis.
  • Potent and orally active multi-target inhibitor of VEGFR and PDGFR family
  • Exhibits prominent antitumor activity
  • Significantly less activity against unrelated RTKs, soluble tyrosine kinases, or serine/threonine kinases
  • Acts as a specific miR-10b inhibitor, blocking miR-10b biogenesis
Catalog No. 50-003-58080 Supplier Medchemexpress LLC Supplier No. HY50751200MG
Linifanib (ABT-869) is a potent, orally active, multi-target inhibitor of the VEGFR and PDGFR family. It demonstrates prominent antitumor activity and has significantly less activity against unrelated RTKs, soluble tyrosine kinases, or serine/threonine kinases. Linifanib also acts as a specific miR-10b inhibitor, blocking miR-10b biogenesis.
  • Potent and orally active multi-target inhibitor of VEGFR and PDGFR family
  • Exhibits prominent antitumor activity
  • Significantly less activity against unrelated RTKs, soluble tyrosine kinases, or serine/threonine kinases
  • Acts as a specific miR-10b inhibitor, blocking miR-10b biogenesis