Learn More
Apexbio Technology LLC Linifanib (ABT-869) 796967-16-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062931
Encompass_Preferred
Linifanib (ABT-869 CAS 796967-16-3) is an ATP-competitive inhibitor of receptor tyrosine kinases targeting platelet-derived growth factor (PDGF) and vascular endothelial growth factor receptors (VEGFRs) as well as FMS-like tyrosine kinase 3 (FLT3) including its activating internal tandem duplication (ITD) mutations Linifanib selectively reduces cell growth in FLT3 ITD-expressing cell lines such as Ba/F3 FLT3 ITD cells inhibits phosphorylation of FLT3 and Akt and induces apoptosis preferentially in ITD mutant cells In vivo experiments with ITD-expressing leukemia xenografts show reduced leukemia progression and prolonged survival in mice treated orally with linifanib This compound is widely used in preclinical research for acute myeloid leukemia studies
Catalog No. 50-000-62931 Supplier Apexbio Technology LLC Supplier No. A29495.1
 Restricted Item
Add to Cart
Add to Cart
Linifanib (ABT-869 CAS 796967-16-3) is an ATP-competitive inhibitor of receptor tyrosine kinases targeting platelet-derived growth factor (PDGF) and vascular endothelial growth factor receptors (VEGFRs) as well as FMS-like tyrosine kinase 3 (FLT3) including its activating internal tandem duplication (ITD) mutations Linifanib selectively reduces cell growth in FLT3 ITD-expressing cell lines such as Ba/F3 FLT3 ITD cells inhibits phosphorylation of FLT3 and Akt and induces apoptosis preferentially in ITD mutant cells In vivo experiments with ITD-expressing leukemia xenografts show reduced leukemia progression and prolonged survival in mice treated orally with linifanib This compound is widely used in preclinical research for acute myeloid leukemia studies