Learn More
Apexbio Technology LLC Linifanib (ABT-869), 5mg. Cas: 796967-16-3 MFCD: MFCD11840918. VEGFR/PDGFR inhibitor
SDP
Supplier:  Apexbio Technology LLC A29495
Encompass_Preferred
Linifanib (ABT-869) is an effective ATP-competitive tyrosine kinase inhibitor against the platelet-derived growth factor (PDGF) receptor and the vascular endothelial growth factor receptor (VEGFR) families, including constitutively active FMS-like receptor tyrosine kinase 3 (FLT3) [1][2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4766