Learn More
Electron Microscopy Sciences COVAMAT Covamat Lining 15 FT
SDP
Catalog No. 501899956
Encompass_Preferred
Clear polyethylene ribbed matting can be used for lining laboratory bench top, shelves, trays, etc. It is easily trimmed and also works well under drums or carboys. 0.010" width 0.030" thick ribs, 30" wide
Catalog No. 50-189-9956 Supplier Electron Microscopy Sciences Supplier No. 7261515
Clear polyethylene ribbed matting can be used for lining laboratory bench top, shelves, trays, etc. It is easily trimmed and also works well under drums or carboys. 0.010" width 0.030" thick ribs, 30" wide