Learn More
Stoelting™ Link 7 Gas Anesthesia System, Mobile with Induction Chamber
SDP
Catalog No. 10000938 Shop All Stoelting Co Products
Change view
Click to view available options
Includes:
Induction chamber
Catalog No. Includes
10-000-938 Induction chamber
1 options
Catalog No. 10-000-938 Supplier Stoelting™ Supplier No. 53931
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Complete Anesthesia System was designed specifically for stereotaxic surgery. System includes a tabletop and/or mobile cart with Isoflurane Vaporizer, Active Gas Scavenger and continuous flow Induction Chamber.
Inhalant Anesthesia System for Stereotaxic Surgery with Active Gas Waste Removal Complete system includes:
  • Tabletop and/or Mobile Anesthesia System Complete with Isoflurane Vaporizer
  • Active Gas Scavenger
  • Continuous Induction Chamber
Features:
  • No flowmeters to adjust. Flip a toggle switch to on to start flow of fresh gas, indicator confirms flow
  • 3 stations for anesthesia with preset flow rate for any station (500cc-5 LPM)
  • Color coded tubing and connector to help user identify which fresh gas supply tubing goes to which station
  • Pressure gauge to let user know when pressure has been released from system for filling of vaporizer.
  • Generates a 50 LPM negative flow that can be attenuated to appropriate levels and divided between 3 stations.
  • Direct vacuum port for use with Vapor-Vac Hooded Induction Chamber
  • Adjustable negative flow meters easily activated with toggle switches.
  • Waste anesthetic gases can be discharged into activated charcoal canister or any non re-circulating hood, vent, downdraft/ back draft system or snorkel.
  • Available without negative flow generating pump- can then be used to attenuate active, in-house vacuum source.
  • Designed for use with the Posi-vac Non-Rebreathing system, Vapor-Vac hooded induction chamber, Stereotaxic nose cone or SCOOP, 3EX Vaporizer. (not included)
  • Higher Throughput
  • Intuitive system- easy to operate, precise, safe flow rate for fresh gas
    Healthy for subjects- less morbidity, less mortality
Specifications
Includes Induction chamber