Learn More
Grainger Siemens SIRIUS Link Module, For 3RV2.1, 3RV2.2, 3RT2.2 Contactors, 1 ea
Catalog No. 502771812
Encompass_Preferred
This link module provides an electrical and mechanical connection for specific contactors, soft starters, and solid-state contactors. It is designed for use with DC operated devices to ensure integrated system functionality.
  • Designed for electrical and mechanical connection of 3RV2.1, 3RV2.2, and 3RT2.2 contactors
  • Compatible with 3RW3 soft starters and 3RF34 solid-state contactors
  • Features screw terminal connections for reliable installation
  • Supports S00 and S0 size circuit breakers and S0 size contactors
  • Three-pole design for versatile application
  • Features screw fixing fastening method
Catalog No. 50-277-1812 Supplier Grainger Supplier No. 13Y508
This link module provides an electrical and mechanical connection for specific contactors, soft starters, and solid-state contactors. It is designed for use with DC operated devices to ensure integrated system functionality.
  • Designed for electrical and mechanical connection of 3RV2.1, 3RV2.2, and 3RT2.2 contactors
  • Compatible with 3RW3 soft starters and 3RF34 solid-state contactors
  • Features screw terminal connections for reliable installation
  • Supports S00 and S0 size circuit breakers and S0 size contactors
  • Three-pole design for versatile application
  • Features screw fixing fastening method