Learn More
Grainger Sirius Link Module, S0 Frame, Screw Terminal, 1 ea
Catalog No. 502773078
Encompass_Preferred
This link module provides reliable electrical and mechanical connection for circuit-breaker and contactor assemblies. Designed for use with S00 and S0 size motor starter protectors and S0 size contactors, it facilitates secure integration in industrial control systems with AC operation.
  • Designed for screw terminal electrical and mechanical connections
  • Compatible with size S00 and S0 motor starter protectors and size S0 contactors
  • Supports AC operation
  • Features three-pole design for versatile application
  • Constructed for robust performance and easy installation
Catalog No. 50-277-3078 Supplier Grainger Supplier No. 13Y507
This link module provides reliable electrical and mechanical connection for circuit-breaker and contactor assemblies. Designed for use with S00 and S0 size motor starter protectors and S0 size contactors, it facilitates secure integration in industrial control systems with AC operation.
  • Designed for screw terminal electrical and mechanical connections
  • Compatible with size S00 and S0 motor starter protectors and size S0 contactors
  • Supports AC operation
  • Features three-pole design for versatile application
  • Constructed for robust performance and easy installation