Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Linking STEM to Agarose Gel Electrophoresis Educational Kit
SDPSCI_ED

Catalog No. s29009 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29009 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29009 Supplier Edvotek™ Supplier No. S46
Only null left
Add to Cart
Add to Cart

Requires

Electrophoresis apparatus, power supply, microwave or hot plate.

Help students to learn about application of gel electrophoresis in DNA Fingerprinting, DNA Paternity Testing, Genetics (related to health and well-being), and the detection of Genetically Modified Foods. Edvotek™ Linking STEM to Agarose Gel Electrophoresis kit is supplied with four unique module options.

  • Links STEM concepts using agarose gel electrophoresis
  • Dyes can be separated in agarose gels and students will use core STEM tools to determine band size
  • Utilize critical thinking and reasoning skills
  • For 10 Lab Groups
  • Complete in 45 minutes
  • Level: Beginner

Specifications

Grade 6 to 12
Includes Instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, micro tipped transfer pipettes.
Time Range Complete in 45 minutes
Content And Storage Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.
Class Size 10 Lab Groups
Product Type Linking STEM to Agarose Gel Electrophoresis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.