Learn More
Alkali Scientific LinkLabel BlueTooth Enabled La
SDP
Catalog No. 503028764
Encompass_Preferred
The new LinkLabelBluetooth Label Maker Lab Printer brings a new level of freedom to laboratory labeling The free downloadable LabelProApp allows users to endlessly customize and specialize their labels Use the app to easily change text font positioning size and label length The LinkLabel also has the ability to print batches serial numbers and a variety of barcodes With 12 available label cassettes the customization doesnt end on your cell phone Choose from multiple colors of tape including transparent Three label heights are available for different sized vessels 6mm tapes are great for tissue culture plates 12mm tapes are ideal from microtubes and the extra tall 24mm tapes are ideal for large tubes and flasks Special ultra-low label tapes are also available for storage in -80C environments or for liquid nitrogen applications Includes 1 L9010-18WK casette Compatible with BlueTooth devices Free app allows for limitless customization Hundreds of preloaded scientific symbols
Catalog No. 50-302-8764 Supplier Alkali Scientific Supplier No. L9300
The new LinkLabelBluetooth Label Maker Lab Printer brings a new level of freedom to laboratory labeling The free downloadable LabelProApp allows users to endlessly customize and specialize their labels Use the app to easily change text font positioning size and label length The LinkLabel also has the ability to print batches serial numbers and a variety of barcodes With 12 available label cassettes the customization doesnt end on your cell phone Choose from multiple colors of tape including transparent Three label heights are available for different sized vessels 6mm tapes are great for tissue culture plates 12mm tapes are ideal from microtubes and the extra tall 24mm tapes are ideal for large tubes and flasks Special ultra-low label tapes are also available for storage in -80C environments or for liquid nitrogen applications Includes 1 L9010-18WK casette Compatible with BlueTooth devices Free app allows for limitless customization Hundreds of preloaded scientific symbols