Learn More
Life Science Products 8-TubeStrip With Individually Attached Piercable Optically Clear Flat Caps, 125/pk
SDP
Catalog No. NC1809172
Encompass_Preferred
0.2 ml PCR 8-LinkStrips with individually attached flat caps. Pierceable, optically clear Flat caps. Ideal for real-time PCR (qPCR). Ultra thin, uniform walls and bottoms for effective temperature transfer.
Catalog No. NC1809172 Supplier Life Science Products Supplier No. LS9509
0.2 ml PCR 8-LinkStrips with individually attached flat caps. Pierceable, optically clear Flat caps. Ideal for real-time PCR (qPCR). Ultra thin, uniform walls and bottoms for effective temperature transfer.