Learn More
Cayman Chemical Linoelaidic Acid methyl ester, Purity: ≥95%, CAS: 2566-97-4, 100 mg
SDP
Catalog No. 502936938
Encompass_Preferred
An esterified form of linoelaidic acid; has been found in H. sabdariffa essential oil as well as biodiesel produced from sea mango seed oil
Catalog No. 50-293-6938 Supplier Cayman Chemical Supplier No. 26732100 MG
An esterified form of linoelaidic acid; has been found in H. sabdariffa essential oil as well as biodiesel produced from sea mango seed oil