Learn More
Cayman Chemical LInoelaIdIc AcId 100mg
SDP
Catalog No. 502895083
Encompass_Preferred
Linoelaidic acid is the all-trans fatty acid homolog of linoleic acid. Trans-fatty acids are generally not detected in raw food oils, but constitute up to 25% of the unsaturated fat in partially hydrogenated oils.{5470}
Catalog No. 50-289-5083 Supplier Cayman Chemical Supplier No. 90160100 MG
Linoelaidic acid is the all-trans fatty acid homolog of linoleic acid. Trans-fatty acids are generally not detected in raw food oils, but constitute up to 25% of the unsaturated fat in partially hydrogenated oils.{5470}