Learn More
Cayman Chemical Linoelaidic Acid, Purity: ≥98%, CAS: 506-21-8, 5 g
SDP
Catalog No. 502895763
Encompass_Preferred
Linoelaidic acid is the all-trans fatty acid homolog of linoleic acid. Trans-fatty acids are generally not detected in raw food oils, but constitute up to 25% of the unsaturated fat in partially hydrogenated oils.{5470}
Catalog No. 50-289-5763 Supplier Cayman Chemical Supplier No. 901605 G
Linoelaidic acid is the all-trans fatty acid homolog of linoleic acid. Trans-fatty acids are generally not detected in raw food oils, but constitute up to 25% of the unsaturated fat in partially hydrogenated oils.{5470}