Learn More
Enzo Life Sciences Linoleamide (50mg). CAS: 3072-13-7
SDP
Supplier:  Enzo Life Sciences BMLFA0230050
Encompass_Preferred
Unsaturated analog of endogenous sleep-inducing lipid. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol or in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-0809