Learn More
Medchemexpress LLC Linoleate sodium | 822-17-3 | 99.88% | 302.43 | 50 MG
SDP
Catalog No. 5000455137
Encompass_Preferred
Linoleate sodium is an orally active IL8 regulator that functions via the JNK and NF-κB pathways. It can modify the composition of fatty acids and the production of metabolites within cells. It exhibits several beneficial activities, including anti-inflammatory, immune-regulating, and tumor cell growth-affecting properties. It is intended for research use only and is not sold to patients.
  • Orally active IL8 regulator.
  • Modulates JNK and NF-κB pathways.
  • Capable of altering fatty acid composition and metabolite production in cells.
  • Possesses anti-inflammatory activity.
  • Offers immune-regulating effects.
  • Affects tumor cell growth.
Catalog No. 50-004-55137 Supplier Medchemexpress LLC Supplier No. HYW01139850MG
Linoleate sodium is an orally active IL8 regulator that functions via the JNK and NF-κB pathways. It can modify the composition of fatty acids and the production of metabolites within cells. It exhibits several beneficial activities, including anti-inflammatory, immune-regulating, and tumor cell growth-affecting properties. It is intended for research use only and is not sold to patients.
  • Orally active IL8 regulator.
  • Modulates JNK and NF-κB pathways.
  • Capable of altering fatty acid composition and metabolite production in cells.
  • Possesses anti-inflammatory activity.
  • Offers immune-regulating effects.
  • Affects tumor cell growth.