Learn More
Medchemexpress LLC Linoleate sodium | 822-17-3 | 99.9% | 302.43 | 100 MG
SDP
Catalog No. 5000455014
Encompass_Preferred
Linoleate sodium is an orally active IL8 regulator that works via the JNK and NF-κB pathway. It can alter the composition of fatty acids and the production of metabolites in cells. Linoleate sodium exhibits anti-inflammatory, immune-regulating, and tumor cell growth-affecting activities.
  • Functions via JNK and NF-κB pathway.
  • Alters the composition of fatty acids.
  • Affects the production of metabolites in cells.
  • Exhibits anti-inflammatory activities.
  • Exhibits immune-regulating activities.
  • Exhibits tumor cell growth-affecting activities.
Catalog No. 50-004-55014 Supplier Medchemexpress LLC Supplier No. HYW011398100MG
Linoleate sodium is an orally active IL8 regulator that works via the JNK and NF-κB pathway. It can alter the composition of fatty acids and the production of metabolites in cells. Linoleate sodium exhibits anti-inflammatory, immune-regulating, and tumor cell growth-affecting activities.
  • Functions via JNK and NF-κB pathway.
  • Alters the composition of fatty acids.
  • Affects the production of metabolites in cells.
  • Exhibits anti-inflammatory activities.
  • Exhibits immune-regulating activities.
  • Exhibits tumor cell growth-affecting activities.