Learn More
Medchemexpress LLC Linoleate sodium | 822-17-3 | 99.9% | 303.44 | 1 G
SDP
Catalog No. 5000453839
Encompass_Preferred
Linoleate sodium is an orally active IL8 regulator that functions via the JNK and NF-κB pathway. It can alter the composition of fatty acids and the production of metabolites within cells. This compound exhibits anti-inflammatory, immune-regulating, and tumor cell growth-affecting activities. It is intended for research use only.
  • Regulates IL8 via JNK and NF-κB pathway
  • Alters fatty acid composition and metabolite production
  • Exhibits anti-inflammatory activity
  • Provides immune-regulating effects
  • Affects tumor cell growth
Catalog No. 50-004-53839 Supplier Medchemexpress LLC Supplier No. HYW0113981G
Linoleate sodium is an orally active IL8 regulator that functions via the JNK and NF-κB pathway. It can alter the composition of fatty acids and the production of metabolites within cells. This compound exhibits anti-inflammatory, immune-regulating, and tumor cell growth-affecting activities. It is intended for research use only.
  • Regulates IL8 via JNK and NF-κB pathway
  • Alters fatty acid composition and metabolite production
  • Exhibits anti-inflammatory activity
  • Provides immune-regulating effects
  • Affects tumor cell growth