Learn More
Cayman Chemical Linoleic Acid glycidyl ester, Purity: ≥98%, CAS: 24305-63-3, 10 mg
SDP
Catalog No. 502909789
Encompass_Preferred
An esterified form of linoleic acid; hydrolyzed to the rat-carcinogenic compound glycidol in vivo; has been found in commercial edible oils and infant formulas
Catalog No. 50-290-9789 Supplier Cayman Chemical Supplier No. 3684010 MG
An esterified form of linoleic acid; hydrolyzed to the rat-carcinogenic compound glycidol in vivo; has been found in commercial edible oils and infant formulas