Learn More
Cayman Chemical LInoleIc ACDd11 ethyl STR
SDP
Catalog No. 502916029
Encompass_Preferred
An internal standard for the quantification of linoleic acid ethyl ester by GC- or LC-MS
Catalog No. 50-291-6029 Supplier Cayman Chemical Supplier No. 37973250 UG
An internal standard for the quantification of linoleic acid ethyl ester by GC- or LC-MS