Learn More
Cayman Chemical LInoleIc ACDd11 ethyl STR
SDP
Supplier:  Cayman Chemical 37973500 UG
Encompass_Preferred
An internal standard for the quantification of linoleic acid ethyl ester by GC- or LC-MS
Catalog No. 50-291-7410