Learn More
Cayman Chemical LInoleIc ACDd11 methyl Ste
SDP
Catalog No. 502988406
Encompass_Preferred
An internal standard for the quantification of linoleic acid methyl ester by GC- or LC-MS
Catalog No. 50-298-8406 Supplier Cayman Chemical Supplier No. 31289100 UG
An internal standard for the quantification of linoleic acid methyl ester by GC- or LC-MS